accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76089-77-5

IUPAC Name: cerium(3+) tritrifluoromethanesulfonate
Synonyms: cerium(3+) tritriflate
Molecular Weight (g/mol): 587.31
Molecular Formula: C3CeF9O9S3
InChi Key: PHSMPGGNMIPKTH-UHFFFAOYSA-K
SMILES: [Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results