accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3COHNH2

CAS RN 760990-10-1

IUPAC Name: 2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 235.09
Molecular Formula: C12H18BNO3
InChi Key: RXHWPAXQYXTPJB-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(N)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results