accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 761441-54-7

LiNNNFFFN

CAS RN 761441-54-7

IUPAC Name: lithium(1+) 4,5-dicyano-2-(trifluoromethyl)-1H-imidazol-1-ide
Synonyms: lithium(1+) 4,5-dicyano-2-(trifluoromethyl)-1H-imidazol-1-ide
Molecular Weight (g/mol): 192.03
Molecular Formula: C6F3LiN4
InChi Key: CVVIFWCYVZRQIY-UHFFFAOYSA-N
SMILES: [Li+].FC(F)(F)C1=NC(C#N)=C([N-]1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results