accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFNH2NH2

CAS RN 76179-40-3

IUPAC Name: 4,5-difluorobenzene-1,2-diamine
Synonyms: 4,5-difluorobenzene-1,2-diamine
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H6F2N2
InChi Key: PPWRHKISAQTCCG-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(F)C=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results