accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 762240-92-6

IUPAC Name: 3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Synonyms: 3-(trifluoromethyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
Molecular Weight (g/mol): 228.6
Molecular Formula: C6H8ClF3N4
InChi Key: AQCSCRYRCRORET-UHFFFAOYSA-N
SMILES: Cl.FC(F)(F)C1=NN=C2CNCCN12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results