accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76283-09-5

FBrBr

CAS RN 76283-09-5

IUPAC Name: 4-bromo-1-(bromomethyl)-2-fluorobenzene
Synonyms: 4-bromo-1-(bromomethyl)-2-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: XMHNLZXYPAULDF-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC=C1CBr
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)