CAS RN 7631-86-9
Thermo Scientific Chemicals Silica gel, for chromatography, 0.035-0.070 mm, 60 A
CAS: 7631-86-9
Silica Gel (2.5-6.0mm) Self Indicating (Iron Salt), Pure, Dessicant Agent, Fisher Chemical™
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Silica gel desiccant, indicating, -6+16 mesh granules
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Thermo Scientific Chemicals Silica gel, for column chrom., ultra pure, 35-70 µm, 60A
CAS: 7631-86-9
Silica gel 60, 0.060-0.2mm (70-230 mesh)
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Thermo Scientific Chemicals Silica gel, for chromatography, 0.200-0.500 mm, 60 A
CAS: 7631-86-9
Silicon(IV) oxide, powder, 1.5 micron, 99.9%
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: silica,silicon dioxide,quartz,cristobalite,diatomaceous earth,tridymite,silicic anhydride,aerosil,infusorial earth,silica, amorphous PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Silica gel, for column chrom., ultrapure, 60-200 micron, 60A
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Silicon dioxide PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Silica gel desiccant, indicating, ACS
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: silica,silicon dioxide,quartz,cristobalite,diatomaceous earth,tridymite,silicic anhydride,aerosil,infusorial earth,silica, amorphous PubChem CID: 24261 ChEBI: CHEBI:30563 IUPAC Name: dioxosilane SMILES: O=[Si]=O
Thermo Scientific Chemicals Silica gel, for chromatography, 0.060-0.200 mm, 60 A
CAS: 7631-86-9
Silica gel (2.5-6.0mm) self indicating, dessicant agent, Fisher Chemical™
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: Silicon dioxide PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
CAS: 7631-86-9 Molecular Formula: O2Si Molecular Weight (g/mol): 60.08 MDL Number: MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 InChI Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N Synonym: silica,silicon dioxide,quartz,cristobalite,diatomaceous earth,tridymite,silicic anhydride,aerosil,infusorial earth,silica, amorphous PubChem CID: 24261 ChEBI: CHEBI:30563 SMILES: O=[Si]=O