accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOHSOO

CAS RN 7631-90-5

IUPAC Name: sodium sulfite
Synonyms: monosodium sulfite
Molecular Weight (g/mol): 103.05
Molecular Formula: NaO3S
InChi Key: DWAQJAXMDSEUJJ-UHFFFAOYSA-L
SMILES: [Na+].[O-]S([O-])=O
Molecular Weight (g/mol) 
  • (14)
  • (4)
Quantity 
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
Grade 
  • (1)
  • (8)
  • (3)
Physical Form 
  • (6)
  • (1)
Percent Purity 
  • (4)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (7)
  • (2)
  • (4)
  • (3)
  • (1)
  • (14)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (8)
  • (3)
  • (6)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)