accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNOOOH

CAS RN 763109-71-3

IUPAC Name: 5-(thiophen-2-yl)-1,2-oxazole-3-carboxylic acid
Synonyms: 5-(thiophen-2-yl)-1,2-oxazole-3-carboxylic acid
Molecular Weight (g/mol): 195.19
Molecular Formula: C8H5NO3S
InChi Key: OBTRHBQUXRJHJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=NOC(=C1)C1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)