accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 763113-22-0

IUPAC Name: 4-{[3-(4-cyclopropanecarbonylpiperazine-1-carbonyl)-4-fluorophenyl]methyl}-1,2-dihydrophthalazin-1-one
Synonyms: olaparib
Molecular Weight (g/mol): 434.47
Molecular Formula: C24H23FN4O3
InChi Key: FDLYAMZZIXQODN-UHFFFAOYSA-N
SMILES: FC1=C(C=C(CC2=NNC(=O)C3=CC=CC=C23)C=C1)C(=O)N1CCN(CC1)C(=O)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results