accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7632-51-1

ClClClClV

CAS RN 7632-51-1

IUPAC Name: vanadium(4+) tetrachloride
Synonyms: vanadium tetrachloride
Molecular Weight (g/mol): 192.74
Molecular Formula: Cl4V
InChi Key: JTJFQBNJBPPZRI-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[V+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results