CAS RN 76335-62-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1’N-Benzyl Biotin, TRC
CAS: 76335-62-1 Molecular Formula: C17H22N2O3S Molecular Weight (g/mol): 334.43 Synonym: (3aS,4S,6aR)-Hexahydro-2-oxo-1-(phenylmethyl)-1H-thieno[3,4-d]imidazole-4-pentanoic Acid,1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-1-(phenylmethyl)-, [3aS-(3aα,4β,6aα)]-,Thieno[3,4-d]imidazoline-4-valeric acid, 1-benzyltetrahydro-2-oxo- (7CI),5-[(3aS,4S,6aR)-3-Benzyl-2-oxohexahydrothieno[3,4-d]imidazol-4-yl]pentanoic Acid IUPAC Name: 5-[(3aR,6S,6aS)-3-benzyl-2-oxo-3a,4,6,6a-tetrahydro-1H-thieno[3,4-d]imidazol-6-yl]pentanoic acid SMILES: OC(=O)CCCC[C@@H]1SC[C@H]2[C@@H]1NC(=O)N2Cc3ccccc3