accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FBFF

CAS RN 7637-07-2

IUPAC Name: trifluoroborane
Synonyms: boron trifluoride boron fluoride bortrifluorid
Molecular Weight (g/mol): 67.81
Molecular Formula: BF3
InChi Key: WTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILES: FB(F)F
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results