accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7646-79-9

IUPAC Name: λ²-cobalt(2+) dichloride
Synonyms: cobalt chloride kobaltdichlorid cobaltous chloride anhydrous cobaltous dichloride cobaltous chloride kobalt chlorid kobalt(II)-chlorid cobalt dichloride dichlorocobalt CoCl2
Molecular Weight (g/mol): 129.83
Molecular Formula: Cl2Co
InChi Key: GVPFVAHMJGGAJG-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Co++]
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (21)
  • (3)
Percent Purity 
  • (2)
  • (9)
  • (1)
Physical Form 
  • (3)
  • (8)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (11)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (5)
  • (2)
  • (3)
  • (21)
  • (3)
  • (2)
  • (9)
  • (1)
  • (3)
  • (8)
  • (1)