accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHSbH3HClClHHClHCl

CAS RN 7647-18-9

IUPAC Name: antimony pentachloride
Synonyms: antimony pentachloride
Molecular Weight (g/mol): 307.07
Molecular Formula: Cl5H8Sb
InChi Key: QFJJIKGZKLCYFT-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.Cl.Cl.[SbH3]
Physical Form 
  • (10)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)