accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76475-62-2

IUPAC Name: 4-(3-carboxypropanamido)benzoic acid
Synonyms: 4-(3-carboxypropanamido)benzoic acid
Molecular Weight (g/mol): 237.21
Molecular Formula: C11H11NO5
InChi Key: IOKXKKSBNJCKOY-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)NC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results