accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2

CAS RN 765-30-0

IUPAC Name: cyclopropanamine
Synonyms: aminocyclopropane cyclopropylamine, 3H-labeled 3h-labeled cyclopropylamine 3H-labeled cyclopropylamine cyclopropylamine, 3h-labeled
Molecular Weight (g/mol): 57.1
Molecular Formula: C3H7N
InChi Key: HTJDQJBWANPRPF-UHFFFAOYSA-N
SMILES: NC1CC1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (4)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (4)
  • (3)
  • (3)