accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(S)H2N

CAS RN 765-38-8

IUPAC Name: (2S)-2-methylpyrrolidin-1-ium
Synonyms: (2S)-2-methylpyrrolidin-1-ium
Molecular Weight (g/mol): 86.16
Molecular Formula: C5H12N
InChi Key: RGHPCLZJAFCTIK-YFKPBYRVSA-O
SMILES: C[C@H]1CCC[NH2+]1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)