accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO

CAS RN 765-70-8

IUPAC Name: 3-methylcyclopentane-1,2-dione
Synonyms: cyclotene
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: OACYKCIZDVVNJL-UHFFFAOYNA-N
SMILES: CC1CCC(=O)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)