accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 765-85-5

IUPAC Name: methyl cyclobutanecarboxylate
Synonyms: methyl cyclobutanecarboxylate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: CBTGNLZUIZHUHY-UHFFFAOYSA-N
SMILES: COC(=O)C1CCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results