accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7651-81-2

IUPAC Name: 2,3-dihydroisoquinolin-3-one
Synonyms: 2H-isoquinolin-3-one
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: GYPOFOQUZZUVQL-UHFFFAOYSA-N
SMILES: O=C1NC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results