accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7651-83-4

IUPAC Name: isoquinolin-7-ol
Synonyms: isoquinolin-7-ol
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: WCRKBMABEPCYII-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=CN=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results