CAS RN 7651-86-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Phenanthrol, TRC
CAS: 7651-86-7 Molecular Formula: C14 H10 O Molecular Weight (g/mol): 194.23 Synonym: 4-Phenanthrenol (9CI, ACI),4-Phenanthrol (6CI, 7CI, 8CI),4-Hydroxyphenanthrene,NSC 171284 IUPAC Name: phenanthren-4-ol SMILES: Oc1cccc2ccc3ccccc3c12