accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76541-44-1

IUPAC Name: 3-(difluoromethyl)pyridine
Synonyms: 3-(difluoromethyl)pyridine
Molecular Weight (g/mol): 129.11
Molecular Formula: C6H5F2N
InChi Key: HKRKGTFBASUFRO-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results