accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHH2N

CAS RN 7658-80-2

IUPAC Name: 2-methylbenzohydrazide
Synonyms: benzoylhydrazine, O-methyl
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: KFXLXEQCRFGDRU-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results