accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CHBr

CAS RN 766-46-1

IUPAC Name: 1-bromo-2-ethynylbenzene
Synonyms: 1-bromo-2-ethynylbenzene
Molecular Weight (g/mol): 181.03
Molecular Formula: C8H5Br
InChi Key: RVDOYUFNRDGYGU-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C#C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)