accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 766-55-2

IUPAC Name: imidazo[1,2-b]pyridazine
Synonyms: imidazo[1,2-b]pyridazine
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: VTVRXITWWZGKHV-UHFFFAOYSA-N
SMILES: C1=CN2N=CC=CC2=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results