accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiCH3

CAS RN 766-77-8

IUPAC Name: dimethyl(phenyl)silyl
Synonyms: dimethyl(phenyl)silyl
Molecular Weight (g/mol): 135.26
Molecular Formula: C8H11Si
InChi Key: OIKHZBFJHONJJB-UHFFFAOYSA-N
SMILES: C[Si](C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)