accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CONH2

CAS RN 7664-66-6

IUPAC Name: 4-(propan-2-yloxy)aniline
Synonyms: 4-isopropoxyaniline
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: MLNFMFAMNBGAQT-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)