CAS RN 7664-93-9
Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution 0.05M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, 96%, extra pure, solution in water
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid, Pure, 50% (w/v), for formaldehyde Determination, Fisher Chemical
Sulfuric Acid Solution ≈5M (10N), Pure
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution 0.25M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution 0.1M (0.2N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid 95%, Certified, for Analysis with Low Cadmium, Lead And Mercury Level, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid 95-98% d=1.83, Primar Plus™, for Trace Metal Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution 0.01M (0.02N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid (Optima™), Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O