accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7669-54-7

IUPAC Name: [(2-nitrophenyl)sulfanyl]chlorane
Synonyms: [(2-nitrophenyl)sulfanyl]chlorane
Molecular Weight (g/mol): 189.61
Molecular Formula: C6H4ClNO2S
InChi Key: NTNKNFHIAFDCSJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1SCl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results