accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NH2NN

CAS RN 767-15-7

IUPAC Name: 4,6-dimethylpyrimidin-2-amine
Synonyms: 2-amino-4,6-dimethylpyrimidine
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: IDQNBVFPZMCDDN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(N)=N1
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results