accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 767-79-3

FNFFFN

CAS RN 767-79-3

IUPAC Name: tetrafluoropyrimidine
Synonyms: tetrafluoropyrimidine
Molecular Weight (g/mol): 152.05
Molecular Formula: C4F4N2
InChi Key: KZMWBUVUQLGBBP-UHFFFAOYSA-N
SMILES: FC1=NC(F)=C(F)C(F)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)