accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiCH3CH3N

CAS RN 7677-24-9

IUPAC Name: trimethylsilanecarbonitrile
Synonyms: trimethylsilyl cyanide
Molecular Weight (g/mol): 99.21
Molecular Formula: C4H9NSi
InChi Key: LEIMLDGFXIOXMT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#N
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results