accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFF(R)H2N

CAS RN 768-31-0

IUPAC Name: (3R)-3-(trifluoromethyl)piperidin-1-ium
Synonyms: (3R)-3-(trifluoromethyl)piperidin-1-ium
Molecular Weight (g/mol): 154.16
Molecular Formula: C6H11F3N
InChi Key: JOHFJTBDUSVGQB-RXMQYKEDSA-O
SMILES: FC(F)(F)[C@@H]1CCC[NH2+]C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)