accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 768-90-1

IUPAC Name: 1-bromoadamantane
Synonyms: 1-bromoadamantane
Molecular Weight (g/mol): 215.13
Molecular Formula: C10H15Br
InChi Key: VQHPRVYDKRESCL-UHFFFAOYSA-N
SMILES: BrC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)