accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7681-38-1

NaOHSOOO

CAS RN 7681-38-1

IUPAC Name: sodium hydrogen sulfate
Synonyms: sodium bisulfate
Molecular Weight (g/mol): 120.05
Molecular Formula: HNaO4S
InChi Key: WBHQBSYUUJJSRZ-UHFFFAOYSA-M
SMILES: [Na+].OS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (9)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)