accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7689-50-1

ClHOONH

CAS RN 7689-50-1

IUPAC Name: 2-(benzylamino)acetic acid hydrochloridyl
Synonyms: (benzylamino)acetic acid hydrochloridyl
Molecular Weight (g/mol): 200.64
Molecular Formula: C9H11ClNO2
InChi Key: YHWVJZULKHCKCK-UHFFFAOYSA-N
SMILES: [Cl].OC(=O)CNCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)