accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3OClHCl

CAS RN 769123-68-4

IUPAC Name: {3-[4-(chloromethyl)phenoxy]propyl}dimethylamine hydrochloride
Synonyms: {3-[4-(chloromethyl)phenoxy]propyl}dimethylamine hydrochloride
Molecular Weight (g/mol): 264.19
Molecular Formula: C12H19Cl2NO
InChi Key: QJUMEHQGZPKXLM-UHFFFAOYSA-N
SMILES: Cl.CN(C)CCCOC1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results