accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76931-93-6

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-(acetylsulfanyl)acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-(acetylsulfanyl)acetate
Molecular Weight (g/mol): 231.22
Molecular Formula: C8H9NO5S
InChi Key: FLCQLSRLQIPNLM-UHFFFAOYSA-N
SMILES: CC(=O)SCC(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results