accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 76985-09-6

NH3(R)OO

CAS RN 76985-09-6

IUPAC Name: (2R)-2-azaniumyl-3-(naphthalen-2-yl)propanoate
Synonyms: (2R)-2-ammonio-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol): 215.25
Molecular Formula: C13H13NO2
InChi Key: JPZXHKDZASGCLU-GFCCVEGCSA-N
SMILES: [NH3+][C@H](CC1=CC=C2C=CC=CC2=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)