accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClH3CNHHNCH3

CAS RN 7699-31-2

IUPAC Name: dihydrogen 1,2-diethylhydrazine dichloride
Synonyms: dihydrogen N,N'-diethylhydrazine dichloride
Molecular Weight (g/mol): 161.07
Molecular Formula: C4H14Cl2N2
InChi Key: NMTDWUVFGOBBAX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCNNCC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results