accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ZnBrBr

CAS RN 7699-45-8

IUPAC Name: zinc(2+) dibromide
Synonyms: zinc bromide
Molecular Weight (g/mol): 225.19
Molecular Formula: Br2Zn
InChi Key: VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES: [Zn++].[Br-].[Br-]
Molecular Weight (g/mol) 
  • (18)
  • (10)
Quantity 
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (5)
  • (1)
  • (5)
  • (3)
  • (9)
Physical Form 
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (1)
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (3)
  • (18)
  • (10)
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (5)
  • (1)
  • (5)
  • (3)
  • (9)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (6)