accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-25-8

CH3OOCH3H3COOH3C

CAS RN 77-25-8

IUPAC Name: 1,3-diethyl 2,2-diethylpropanedioate
Synonyms: 1,3-diethyl 2,2-diethylpropanedioate
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: ZKBBUZRGPULIRN-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC)(CC)C(=O)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)