accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-36-1

IUPAC Name: 2-chloro-5-(1-hydroxy-3-oxo-2,3-dihydro-1H-isoindol-1-yl)benzene-1-sulfonamide
Synonyms: chlorthalidone oksodolin (oxodolin) igroton chlortalidone (inn) (+-)-chlorthalidone oxodoline chlorphthalidolone zambesil phthalamodine hydro-long
Molecular Weight (g/mol): 338.76
Molecular Formula: C14H11ClN2O4S
InChi Key: JIVPVXMEBJLZRO-UHFFFAOYNA-N
SMILES: NS(=O)(=O)C1=CC(=CC=C1Cl)C1(O)NC(=O)C2=CC=CC=C12
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results