accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClClClCl

CAS RN 77-47-4

IUPAC Name: hexachlorocyclopenta-1,3-diene
Synonyms: hexachlorocyclopentadiene
Molecular Weight (g/mol): 272.76
Molecular Formula: C5Cl6
InChi Key: VUNCWTMEJYMOOR-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(Cl)(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results