accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-48-5

IUPAC Name: 1,3-dibromo-5,5-dimethylimidazolidine-2,4-dione
Synonyms: dbdmh
Molecular Weight (g/mol): 285.92
Molecular Formula: C5H6Br2N2O2
InChi Key: VRLDVERQJMEPIF-UHFFFAOYSA-N
SMILES: CC1(C)N(Br)C(=O)N(Br)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results