accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOO

CAS RN 77-55-4

IUPAC Name: 1-phenylcyclopentane-1-carboxylic acid
Synonyms: 1-phenylcyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: RHPCYZLXNNRRMB-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCCC1)C1=CC=CC=C1