accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COHNO

CAS RN 77-67-8

IUPAC Name: 3-ethyl-3-methylpyrrolidine-2,5-dione
Synonyms: ethosuximide etosuximida aethosuximide
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: HAPOVYFOVVWLRS-UHFFFAOYNA-N
SMILES: CCC1(C)CC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)