accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 77-93-0

IUPAC Name: 1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: triethyl citrate citroflex 2 hydragen cat triaethylcitrat triethylester kyseliny citronove eudraflex
Molecular Weight (g/mol): 276.29
Molecular Formula: C12H20O7
InChi Key: DOOTYTYQINUNNV-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results